Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenformin hydrochloride | 834-28-6 | 99.6% | 241.72 | 5 G
SDP

Catalog No. 5000391205
Encompass_Preferred

Phenformin hydrochloride is an anti-diabetic agent from the biguanide class that can activate AMPK activity. It is intended for research use only and is a white to off-white solid.

  • Activates AMPK activity
  • Solid appearance
  • White to off-white color
  • Store at 4°C under nitrogen, away from moisture
  • Soluble in H2O and DMSO

Catalog No. 50-003-91205 Supplier Medchemexpress LLC Supplier No. HY16397A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Phenformin hydrochloride is an anti-diabetic agent from the biguanide class that can activate AMPK activity. It is intended for research use only and is a white to off-white solid.

  • Activates AMPK activity
  • Solid appearance
  • White to off-white color
  • Store at 4°C under nitrogen, away from moisture
  • Soluble in H2O and DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.