Learn More
Medchemexpress LLC Phenibut | 1078-21-3 | 99.9% | 179.22 | 250 MG
SDP
Catalog No. 5000347339
Encompass_Preferred
Phenibut (β-Phenyl-GABA) is a GABA-B agonist. It acts as a GABA-mimetic, primarily at GABAB receptors. Phenibut has anxiolytic and nootropic (cognition enhancing) effects. It is also a neuropsychotropic agent and stimulates dopamine receptors, while antagonizing β-phenethylamine (PEA), which is an endogenous anxiogenic.
  • Acts as a GABA-mimetic
  • Has anxiolytic effects
  • Has nootropic (cognition enhancing) effects
  • Neuropsychotropic agent
  • Stimulates dopamine receptors
  • Antagonizes beta-phenethylamine (PEA)
  • Appears as a white to off-white solid
  • Purity of 99.9%
Catalog No. 50-003-47339 Supplier Medchemexpress LLC Supplier No. 250MGHY108403
Phenibut (β-Phenyl-GABA) is a GABA-B agonist. It acts as a GABA-mimetic, primarily at GABAB receptors. Phenibut has anxiolytic and nootropic (cognition enhancing) effects. It is also a neuropsychotropic agent and stimulates dopamine receptors, while antagonizing β-phenethylamine (PEA), which is an endogenous anxiogenic.
  • Acts as a GABA-mimetic
  • Has anxiolytic effects
  • Has nootropic (cognition enhancing) effects
  • Neuropsychotropic agent
  • Stimulates dopamine receptors
  • Antagonizes beta-phenethylamine (PEA)
  • Appears as a white to off-white solid
  • Purity of 99.9%