Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenibut hydrochloride | 3060-41-1 | ≥98.0% | 215.68 | C10H14ClNO2 | 1 ML
SDP

Catalog No. 5000209177
Encompass_Preferred

Phenibut hydrochloride is a GABAB receptor agonist supplied as a 10 mM solution in DMSO for research use. It is provided as a ready-to-use solution intended for in vitro and neuroscience applications.

  • supplied concentration: 10 mM in DMSO
  • volume: 1 mL per vial
  • cas number: 3060-41-1
  • molecular formula: C10H14ClNO2
  • molecular weight: 215.68
  • assay purity (nmr): ≥98.0%
  • storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month)

Catalog No. 50-002-09177 Supplier Medchemexpress LLC Supplier No. HYW04133310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenibut hydrochloride is a GABAB receptor agonist supplied as a 10 mM solution in DMSO for research use. It is provided as a ready-to-use solution intended for in vitro and neuroscience applications.

  • supplied concentration: 10 mM in DMSO
  • volume: 1 mL per vial
  • cas number: 3060-41-1
  • molecular formula: C10H14ClNO2
  • molecular weight: 215.68
  • assay purity (nmr): ≥98.0%
  • storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.