Learn More
Medchemexpress LLC HY-W010144 500mg Medchemexpress, Phenidone CAS:92-43-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW010144500MG
Encompass_Preferred
Medchemexpress, HY-W010144 500mg Phenidone CAS:92-43-3 Phenidone, an orally active dual inhibitor of cyclooxygenase (COX) and lipoxygenase (LOX), ameliorates rat paralysis in experimental autoimmune encephalomyelitis. Phenidone is a potent hypotensive agent in the spontaneously hypertensive rat. Phenidone is used as a photographic developer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3284