Learn More
Medchemexpress LLC Phenindione-d5 | 70711-53-4 | 99.5% | 1 MG
SDP
Catalog No. 5000406591
Encompass_Preferred
Phenindione-d5 is deuterium labeled Phenindione, an anticoagulant that functions as a Vitamin K antagonist. It is for research use only and not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06591 Supplier Medchemexpress LLC Supplier No. HYB0325S1MG
Phenindione-d5 is deuterium labeled Phenindione, an anticoagulant that functions as a Vitamin K antagonist. It is for research use only and not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS