Learn More
Medchemexpress LLC Phenindione-d5 | 70711-53-4 | 99.5% | 5 MG
SDP
Catalog No. 5000406592
Encompass_Preferred
Phenindione-d5 is deuterium labeled Phenindione, which is an anticoagulant that functions as a Vitamin K antagonist.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06592 Supplier Medchemexpress LLC Supplier No. HYB0325S5MG
Phenindione-d5 is deuterium labeled Phenindione, which is an anticoagulant that functions as a Vitamin K antagonist.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.