Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenindione-d5 | 70711-53-4 | MFCD00630829 | 99.5% | 227.27 | C15H5D5O2 | 10 MG
SDP

Catalog No. 5000298235
Encompass_Preferred

Phenindione-d5 is a deuterium-labeled analog of phenindione supplied as a research reagent and internal standard for quantitative analysis. It functions as a vitamin K antagonist and is intended for tracer studies and analytical applications such as LC-MS, GC-MS, and NMR.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity: 99.5%.
  • Physical form: white to off-white solid.
  • Storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Solubility: DMSO ≥100 mg/mL; water 0.67 mg/mL (requires ultrasonic).
  • Packaged amount: 10 mg suitable for analytical use.

Catalog No. 50-002-98235 Supplier Medchemexpress LLC Supplier No. HYB0325S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenindione-d5 is a deuterium-labeled analog of phenindione supplied as a research reagent and internal standard for quantitative analysis. It functions as a vitamin K antagonist and is intended for tracer studies and analytical applications such as LC-MS, GC-MS, and NMR.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity: 99.5%.
  • Physical form: white to off-white solid.
  • Storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Solubility: DMSO ≥100 mg/mL; water 0.67 mg/mL (requires ultrasonic).
  • Packaged amount: 10 mg suitable for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.