Learn More
Medchemexpress LLC Pheniprazine hydrochloride | 66-05-7 | 98.7% | 186.68 g·mol⁻¹ | C9H15ClN2 | 50 MG
SDP
Catalog No. 5000280492
Encompass_Preferred
Pheniprazine hydrochloride is the hydrochloride salt of pheniprazine, an irreversible, nonselective monoamine oxidase (MAOI) inhibitor supplied for research use in biochemical and pharmacological studies of monoamine systems and depression.
  • Irreversible, nonselective monoamine oxidase inhibitor.
  • Useful research tool for studies of depression and monoaminergic signaling.
  • Supplied as a white to off-white solid.
  • Typical purity about 98.7%.
  • Available in small research quantities, for example 50 mg.
  • Store solid at 4°C, sealed and away from moisture; in solution store at -80°C for long-term storage.
Catalog No. 50-002-80492 Supplier Medchemexpress LLC Supplier No. HYW224327A50MG
Pheniprazine hydrochloride is the hydrochloride salt of pheniprazine, an irreversible, nonselective monoamine oxidase (MAOI) inhibitor supplied for research use in biochemical and pharmacological studies of monoamine systems and depression.
  • Irreversible, nonselective monoamine oxidase inhibitor.
  • Useful research tool for studies of depression and monoaminergic signaling.
  • Supplied as a white to off-white solid.
  • Typical purity about 98.7%.
  • Available in small research quantities, for example 50 mg.
  • Store solid at 4°C, sealed and away from moisture; in solution store at -80°C for long-term storage.