Learn More
Cayman Chemical Pheniramine (maleate), Purity: ≥98%, CAS: 132-20-7, 50 mg
SDP
Catalog No. 502917350
Encompass_Preferred
An analytical reference standard categorized as an antihistamine; has antidepressant-like activity in rats; associated with fatal and non-fatal overdoses; intended for research and forensic applications
Catalog No. 50-291-7350 Supplier Cayman Chemical Supplier No. 3947350 MG
An analytical reference standard categorized as an antihistamine; has antidepressant-like activity in rats; associated with fatal and non-fatal overdoses; intended for research and forensic applications