Learn More
Aqua Solutions Phenolphthalein 1% in 95% Alcohol (pH 8.0 Clear, pH 10.0 Pink) (250ML)
SDP
Catalog No. NC0173493
Encompass_Preferred
Phenolphthalein 1% in 95% Alcohol (pH 8.0 Clear, pH 10.0 Pink) (250ML)
Catalog No. NC0173493 Supplier Aqua Solutions Supplier No. P1997250ML
Phenolphthalein 1% in 95% Alcohol (pH 8.0 Clear, pH 10.0 Pink) (250ML)