Learn More
LaMarKa Chemical Company Phenolphthalein Indicator 0.5% W/V (5 gpl) in Ethanol, 0.5l
Catalog No. 504400277
Encompass_Preferred
Phenolphthalein Indicator 0.5% W/V (5 gpl) in Ethanol, 0.5l Plastic Bottle
Catalog No. 50-440-0277 Supplier LaMarKa Chemical Company Supplier No. 00001188167482411
Phenolphthalein Indicator 0.5% W/V (5 gpl) in Ethanol, 0.5l Plastic Bottle