Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenofluor(Tm) Solution

Catalog No. 79529110ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-529-110ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 79-529-110ML Supplier Sigma Aldrich Supplier No. 79529110ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Facile deoxyfluorination of phenols without preactivation (), along with highly chemoselective deoxyfluorination of highly functionalized late-stage intermediates ().PhenoFluor is a trademark of SciFluor Life Sciences, LLC

Specifications

CAS 680-31-9
Melting Point 7°C
Density 1.03 g/mL at 25°C
Boiling Point 230°C to 232°C (740 mmHg, lit.)
Molecular Formula C6H18N3OP
Linear Formula [(CH3)2N]3PO
Quantity 10 mL
MDL Number MFCD00008303
Synonym 1,3-Bis(2,6-diisopropylphenyl)-2,2-difluoro-4-imidazoline
Molecular Weight (g/mol) 179.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.