Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenol-1,2-Dichlorobenzene Mixture

Catalog No. 3352225L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-522-25L 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 33-522-25L Supplier Sigma Aldrich Supplier No. 335222.5L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenol-1,2-dichlorobenzene mixture is a mixture of phenol and 1,2-dichlorobenzene in the ratio of 1:1. 1,2-Dichlorobenzene/Phenol mixture (50/50wt%) has been used as solvent for poly(butylene succinate) (PBS) during the evaluation of intrinsic viscosity of PBS by Huggins and Kramer plots, and from the single-point methods.Phenol-1,2-dichlorobenzene mixture may be used as a solvent to measure the intrinsic viscosity of a polymer sample using an Ubbelohde viscometer.

Specifications

CAS 28322-40-9
Melting Point 41°C to 43°C
Molecular Formula C5H6O3
Quantity 1 g
Molecular Weight (g/mol) 114.1 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.