Learn More
Cayman Chemical Phenol-13C6, Purity: ≥98%, CAS: 89059-34-7, 1 g
SDP
Catalog No. 502989852
Encompass_Preferred
A building block; has been used in the synthesis of 13C6-labeled metabolites of (±)-asenapine
Catalog No. 50-298-9852 Supplier Cayman Chemical Supplier No. 305261 G
A building block; has been used in the synthesis of 13C6-labeled metabolites of (±)-asenapine