Learn More
Cayman Chemical Phenol-13C6 250mg
SDP
Catalog No. 502991145
Encompass_Preferred
A building block; has been used in the synthesis of 13C6-labeled metabolites of (±)-asenapine
Catalog No. 50-299-1145 Supplier Cayman Chemical Supplier No. 30526250 MG
A building block; has been used in the synthesis of 13C6-labeled metabolites of (±)-asenapine