Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 2-(3,5-dimethyl-1H-pyrazol-1-yl)-5-methyl- | 1391758-52-3 | 100.0% | 202.25 | C12H14N2O | 10 MG
SDP

Catalog No. 5000242141
Encompass_Preferred

ME1111 is a high-purity small-molecule antifungal agent that inhibits succinate dehydrogenase in Trichophyton species. It is supplied for laboratory research, with formula C12H14N2O, molecular weight 202.25 g/mol, CAS 1391758-52-3, and notable solubility in DMSO.

  • High purity (99.95%).
  • Molecular weight 202.25 g/mol.
  • Chemical formula C12H14N2O.
  • Soluble in DMSO at 100 mg/mL.
  • Available in small-scale quantities suitable for research.
  • Powder stable at -20°C for long-term storage; solution storage is short term.
  • Intended for in vitro and preclinical research use only.

Catalog No. 50-002-42141 Supplier Medchemexpress LLC Supplier No. HY10801210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ME1111 is a high-purity small-molecule antifungal agent that inhibits succinate dehydrogenase in Trichophyton species. It is supplied for laboratory research, with formula C12H14N2O, molecular weight 202.25 g/mol, CAS 1391758-52-3, and notable solubility in DMSO.

  • High purity (99.95%).
  • Molecular weight 202.25 g/mol.
  • Chemical formula C12H14N2O.
  • Soluble in DMSO at 100 mg/mL.
  • Available in small-scale quantities suitable for research.
  • Powder stable at -20°C for long-term storage; solution storage is short term.
  • Intended for in vitro and preclinical research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.