Learn More
Medchemexpress LLC Phenol, 2,4,6-triiodo- | 609-23-4 | 99.5% | 471.80 g/mol | C6H3I3O | 1 ML
SDP
Catalog No. 5000210498
Encompass_Preferred
2,4,6-triiodophenol is a small-molecule research reagent used as a leukotriene B4 (LTB4) synthesis inhibitor and reported to induce apoptosis in mouse blastocysts. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available in multiple solid sizes for formulation and storage flexibility.
  • Orally active LTB4 synthesis inhibitor, useful for inflammation and leukotriene signaling studies.
  • Reported to induce mouse blastocyst apoptosis, suitable for apoptosis and developmental toxicity research.
  • Supplied as a ready-to-use 10 mM solution in DMSO for immediate experimental use.
  • High chemical purity (99.5%) with analytical data available for quality control.
  • Stable when stored at 4°C protected from light; in solvent, store at -80°C for long-term storage.
Catalog No. 50-002-10498 Supplier Medchemexpress LLC Supplier No. HYW07368710MM/1ML
2,4,6-triiodophenol is a small-molecule research reagent used as a leukotriene B4 (LTB4) synthesis inhibitor and reported to induce apoptosis in mouse blastocysts. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and is also available in multiple solid sizes for formulation and storage flexibility.
  • Orally active LTB4 synthesis inhibitor, useful for inflammation and leukotriene signaling studies.
  • Reported to induce mouse blastocyst apoptosis, suitable for apoptosis and developmental toxicity research.
  • Supplied as a ready-to-use 10 mM solution in DMSO for immediate experimental use.
  • High chemical purity (99.5%) with analytical data available for quality control.
  • Stable when stored at 4°C protected from light; in solvent, store at -80°C for long-term storage.