Learn More
Medchemexpress LLC Phenol, 2,4-bis(1,1-dimethylethyl)- | 96-76-4 | MFCD00008828 | 100.0% | 206.33 g/mol | C14H22O | 500 G
SDP
Catalog No. 5000302635
Encompass_Preferred
2,4-Di-tert-butylphenol (CAS 96-76-4) is a substituted phenol used as a research chemical and chemical intermediate. It has reported antioxidant and anti-inflammatory activities, and is used in studies of nuclear receptor modulation and in the manufacture of antioxidants, UV stabilizers, and related compounds.
  • High purity suitable for analytical and research use.
  • White to yellow solid for easy handling and storage.
  • Exhibits RXRα activation and antioxidant activity in biological assays.
  • Useful intermediate for synthesis of antioxidants and UV stabilizers.
  • Available in bulk quantities upon request for scale-up and production.
Catalog No. 50-003-02635 Supplier Medchemexpress LLC Supplier No. HYW014589500G
2,4-Di-tert-butylphenol (CAS 96-76-4) is a substituted phenol used as a research chemical and chemical intermediate. It has reported antioxidant and anti-inflammatory activities, and is used in studies of nuclear receptor modulation and in the manufacture of antioxidants, UV stabilizers, and related compounds.
  • High purity suitable for analytical and research use.
  • White to yellow solid for easy handling and storage.
  • Exhibits RXRα activation and antioxidant activity in biological assays.
  • Useful intermediate for synthesis of antioxidants and UV stabilizers.
  • Available in bulk quantities upon request for scale-up and production.