Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 2-bromo-6-nitro- | 13073-25-1 | MFCD02683216 | 98.0% | 218.01 g/mol | C6H4BrNO3 | 10 G
SDP

Catalog No. 5000208816
Encompass_Preferred

2-Bromo-6-nitrophenol is a brominated nitrophenol intermediate supplied for laboratory research. It is used in organic synthesis and can be converted via 2-bromo-6-aminophenol to N-acetyl-2-bromo-6-aminophenol. Supplied as a solid for bench-top handling; for research use only.

  • Purity 98.0% as listed on certificate of analysis.
  • Molecular formula C6H4BrNO3 and molecular weight 218.01 g/mol.
  • Appearance: solid, light brown to yellow.
  • Intended for research and synthetic applications.
  • Provided in small-scale package suitable for laboratory use.

Catalog No. 50-002-08816 Supplier Medchemexpress LLC Supplier No. HYW04088610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Bromo-6-nitrophenol is a brominated nitrophenol intermediate supplied for laboratory research. It is used in organic synthesis and can be converted via 2-bromo-6-aminophenol to N-acetyl-2-bromo-6-aminophenol. Supplied as a solid for bench-top handling; for research use only.

  • Purity 98.0% as listed on certificate of analysis.
  • Molecular formula C6H4BrNO3 and molecular weight 218.01 g/mol.
  • Appearance: solid, light brown to yellow.
  • Intended for research and synthetic applications.
  • Provided in small-scale package suitable for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.