Learn More
Medchemexpress LLC Phenol, 2-fluoro- | 367-12-4 | 99.96% | 112.10 | 10 G
SDP
Catalog No. 5000404974
Encompass_Preferred
2-Fluorophenol is a biochemical reagent utilized as a biological material or organic compound for various life science research applications. This product is intended for research use only and is not sold for patient use.
  • Biochemical reagent for life science research
  • Store at 4°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
Catalog No. 50-004-04974 Supplier Medchemexpress LLC Supplier No. HY4175510G
2-Fluorophenol is a biochemical reagent utilized as a biological material or organic compound for various life science research applications. This product is intended for research use only and is not sold for patient use.
  • Biochemical reagent for life science research
  • Store at 4°C under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen