Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 2-(methoxy-d3)- | 74495-69-5 | 99.0% | 127.16 g·mol⁻1 | C7H5D3O2 | 10 MG
SDP

Catalog No. 5000295617
Encompass_Preferred

Guaiacol-d3 (2-Methoxyphenol-d3) is a deuterium-labeled isotopologue of guaiacol supplied as a high-purity analytical standard and research reagent. It is intended for use as a stable isotope internal standard, in mass spectrometry, metabolic tracing, and mechanistic studies where deuterium labeling facilitates compound identification and quantitation.

  • Deuterium-labeled guaiacol for analytical applications.
  • Chemical formula C7H5D3O2; molecular weight 127.16 g·mol⁻1.
  • High purity: 99.0%.
  • Suitable as a mass spectrometry internal standard and tracer.
  • Available in small pack sizes for laboratory use (e.g., 10 MG).

Catalog No. 50-002-95617 Supplier Medchemexpress LLC Supplier No. HYN1380S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Guaiacol-d3 (2-Methoxyphenol-d3) is a deuterium-labeled isotopologue of guaiacol supplied as a high-purity analytical standard and research reagent. It is intended for use as a stable isotope internal standard, in mass spectrometry, metabolic tracing, and mechanistic studies where deuterium labeling facilitates compound identification and quantitation.

  • Deuterium-labeled guaiacol for analytical applications.
  • Chemical formula C7H5D3O2; molecular weight 127.16 g·mol⁻1.
  • High purity: 99.0%.
  • Suitable as a mass spectrometry internal standard and tracer.
  • Available in small pack sizes for laboratory use (e.g., 10 MG).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.