Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenol-23456-d5 98 atom D1G
Catalog No. 502898095
Encompass
Phenol-2 3 4 5 6-d5 is an isotope-labeled analog of phenol wherein 2 3 4 5 and 6th protons of phenol are replaced by deuterium.
Catalog No. 50-289-8095 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4253701G
Phenol-2 3 4 5 6-d5 is an isotope-labeled analog of phenol wherein 2 3 4 5 and 6th protons of phenol are replaced by deuterium.