Learn More
Medchemexpress LLC Phenol, 3,5-bis(trifluoromethyl)- | 349-58-6 | 99.87% | 230.11 | 10 G
SDP
Catalog No. 5000454007
Encompass_Preferred
KG-655 is a small molecule compound that binds to the β-folded surface and internal cavity of ARNT PAS-B and interferes with its binding to the transcriptional coactivator TACC3. It promotes the homodimerization of ARNT PAS-B and has the potential to regulate multiple ARNT-mediated signaling pathways.
  • Binds to the β-folded surface and internal cavity of ARNT PAS-B
  • Interferes with ARNT PAS-B binding to the transcriptional coactivator TACC3
  • Promotes homodimerization of ARNT PAS-B
  • Potential to regulate multiple ARNT-mediated signaling pathways
  • For research use only
Catalog No. 50-004-54007 Supplier Medchemexpress LLC Supplier No. HYW01224810G
KG-655 is a small molecule compound that binds to the β-folded surface and internal cavity of ARNT PAS-B and interferes with its binding to the transcriptional coactivator TACC3. It promotes the homodimerization of ARNT PAS-B and has the potential to regulate multiple ARNT-mediated signaling pathways.
  • Binds to the β-folded surface and internal cavity of ARNT PAS-B
  • Interferes with ARNT PAS-B binding to the transcriptional coactivator TACC3
  • Promotes homodimerization of ARNT PAS-B
  • Potential to regulate multiple ARNT-mediated signaling pathways
  • For research use only