Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 3-[(9H-purin-6-ylamino)methyl]- | 75737-38-1 | 98.0% | 241.25 | 1 ML
SDP

Catalog No. 5000437207
Encompass_Preferred

Meta-topolin (m-Topolin) is a highly active aromatic cytokinin, naturally occurring in plant tissues. It was initially isolated from poplar leaves and is found alongside its 9-β-D-ribofuranosyl and 9-β-v-glucopyranosyl derivatives. This compound demonstrates greater activity than zeatin and benzyladenine in promoting shoot formation within plant tissue cultures, and shows comparable efficacy to BAP in shoot production in *Spathiphyllum floribundum*.

  • Highly active aromatic cytokinin
  • Natural constituent of plant tissues
  • Promotes shoot formation in plant tissue cultures
  • Comparable efficacy to BAP in shoot production
  • Appearance: White to off-white solid
  • Molecular weight: 241.25
  • CAS No.: 75737-38-1

Catalog No. 50-004-37207 Supplier Medchemexpress LLC Supplier No. HY11210410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Meta-topolin (m-Topolin) is a highly active aromatic cytokinin, naturally occurring in plant tissues. It was initially isolated from poplar leaves and is found alongside its 9-β-D-ribofuranosyl and 9-β-v-glucopyranosyl derivatives. This compound demonstrates greater activity than zeatin and benzyladenine in promoting shoot formation within plant tissue cultures, and shows comparable efficacy to BAP in shoot production in *Spathiphyllum floribundum*.

  • Highly active aromatic cytokinin
  • Natural constituent of plant tissues
  • Promotes shoot formation in plant tissue cultures
  • Comparable efficacy to BAP in shoot production
  • Appearance: White to off-white solid
  • Molecular weight: 241.25
  • CAS No.: 75737-38-1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.