Learn More
Medchemexpress LLC Phenol, 4-(1-methyl-1-phenylethyl)- | 599-64-4 | MFCD00002365 | 99.6% | 212.29 g/mol | C15H16O | 500 G
SDP
Catalog No. 5000304143
Encompass_Preferred
4-Cumylphenol (CAS 599-64-4) is a phenolic compound supplied as a white to off-white solid. It is used as a polycarbonate chain terminator and as an intermediate in the production of plastics, surfactants, and fungicides, and is used in biochemical research. The material is supplied with analytical documentation and storage recommendations for laboratory and industrial use.
  • High purity (99.6%).
  • White to off-white solid appearance.
  • Functions as a polycarbonate chain terminator and chemical intermediate.
  • Provided with datasheet, SDS, and certificate of analysis.
  • Recommended storage under nitrogen; cold storage for solutions.
Catalog No. 50-003-04143 Supplier Medchemexpress LLC Supplier No. HYW014282500G
Add to Cart
Add to Cart
4-Cumylphenol (CAS 599-64-4) is a phenolic compound supplied as a white to off-white solid. It is used as a polycarbonate chain terminator and as an intermediate in the production of plastics, surfactants, and fungicides, and is used in biochemical research. The material is supplied with analytical documentation and storage recommendations for laboratory and industrial use.
  • High purity (99.6%).
  • White to off-white solid appearance.
  • Functions as a polycarbonate chain terminator and chemical intermediate.
  • Provided with datasheet, SDS, and certificate of analysis.
  • Recommended storage under nitrogen; cold storage for solutions.