Learn More
Medchemexpress LLC Phenol, 4,4'-(1,1-dioxido-3H-2,1-benzoxathiol-3-ylidene)bis[2-bromo-3-methyl-6-(1-methylethyl)-, sodium | 34722-90-2 | 95.0% | 10 G
SDP
Catalog No. 5000411035
Encompass_Preferred
Bromothymol Blue sodium salt is a pH indicator. It is for research use only and not sold to patients. Protect from light during storage.
  • Ph indicator
Catalog No. 50-004-11035 Supplier Medchemexpress LLC Supplier No. HYD0012A10G
Bromothymol Blue sodium salt is a pH indicator. It is for research use only and not sold to patients. Protect from light during storage.
  • Ph indicator