Learn More
Medchemexpress LLC Phenol, 4,4',4'-(1-methyl-1-propanyl-3-ylidene)tris[2-(1,1-dimethylethyl)-5-methyl- | 1843-03-4 | 98.1% | 544.81 | 10 MG
SDP
Catalog No. 5000437734
Encompass_Preferred
Topanol CA is an effective antioxidant and stabilizer. It is designed for use with resins and light-colored rubber products. This compound is intended for research purposes only.
  • Effective antioxidant
  • Stabilizes resins
  • Stabilizes light-colored rubber products
Catalog No. 50-004-37734 Supplier Medchemexpress LLC Supplier No. HY11279410MG
Topanol CA is an effective antioxidant and stabilizer. It is designed for use with resins and light-colored rubber products. This compound is intended for research purposes only.
  • Effective antioxidant
  • Stabilizes resins
  • Stabilizes light-colored rubber products