Learn More
Medchemexpress LLC Phenol, 4,4′,4′′,4′′′-(21H,23H-porphine-5,10,15,20-tetrayl)tetrakis- | 51094-17-8 | MFCD00191501 | 98.0% | 678.73 g/mol | C44H30N4O4 | 500 MG
SDP
Catalog No. 5000299369
Encompass_Preferred
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin is a meso-substituted porphyrin used as a biochemical reagent in life-science research, photophysical studies, photodynamic research, and materials chemistry. Supplied as a solid with high purity, it is suitable for synthesis, assay development, and photophysical characterization.
  • High purity (98.0%) suitable for analytical and research use.
  • Solid form, easy to handle and store.
  • Molecular weight 678.73 g/mol and formula C44H30N4O4 for stoichiometric calculations.
  • Available in multiple pack sizes to support small- and large-scale experiments.
  • Stable under recommended storage; solution storage guidelines available.
  • Useful in photodynamic, photophysical, and synthetic applications.
Catalog No. 50-002-99369 Supplier Medchemexpress LLC Supplier No. HYW035140500MG
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin is a meso-substituted porphyrin used as a biochemical reagent in life-science research, photophysical studies, photodynamic research, and materials chemistry. Supplied as a solid with high purity, it is suitable for synthesis, assay development, and photophysical characterization.
  • High purity (98.0%) suitable for analytical and research use.
  • Solid form, easy to handle and store.
  • Molecular weight 678.73 g/mol and formula C44H30N4O4 for stoichiometric calculations.
  • Available in multiple pack sizes to support small- and large-scale experiments.
  • Stable under recommended storage; solution storage guidelines available.
  • Useful in photodynamic, photophysical, and synthetic applications.