Learn More
Medchemexpress LLC Phenol, 4-[[4-(4-chlorophenyl)-2-thiazolyl]amino]- | 312636-16-1 | 99.25% | 302.78 | 200 MG
SDP
Catalog No. 5000348621
Encompass_Preferred
SKI-II is an orally active and synthetic inhibitor of sphingosine kinase (SK) activity. It is intended for research use only and causes irreversible inhibition of SK1 by inducing its lysosomal and/or proteasomal degradation. It inhibits cell proliferation and promotes β-catenin degradation.
  • Orally active and synthetic inhibitor of sphingosine kinase (SK) activity
  • IC50 values: 78 μM for SK1 and 45 μM for SK2
  • Causes irreversible inhibition of SK1 by inducing its lysosomal and/or proteasomal degradation
  • Inhibits cell proliferation by suppressing the Wnt/β-catenin signaling pathway
  • Promotes the degradation of β-catenin by enhancing Wnt5A
  • Shows synergistic effect with DDP in human gastric carcinoma SGC7901/DDP cell line
  • Reduces S1P concentrations in plasma in mice with chronic administration
  • Reduces tumor growth in mice bearing solid tumor models
Catalog No. 50-003-48621 Supplier Medchemexpress LLC Supplier No. HY13822200MG
SKI-II is an orally active and synthetic inhibitor of sphingosine kinase (SK) activity. It is intended for research use only and causes irreversible inhibition of SK1 by inducing its lysosomal and/or proteasomal degradation. It inhibits cell proliferation and promotes β-catenin degradation.
  • Orally active and synthetic inhibitor of sphingosine kinase (SK) activity
  • IC50 values: 78 μM for SK1 and 45 μM for SK2
  • Causes irreversible inhibition of SK1 by inducing its lysosomal and/or proteasomal degradation
  • Inhibits cell proliferation by suppressing the Wnt/β-catenin signaling pathway
  • Promotes the degradation of β-catenin by enhancing Wnt5A
  • Shows synergistic effect with DDP in human gastric carcinoma SGC7901/DDP cell line
  • Reduces S1P concentrations in plasma in mice with chronic administration
  • Reduces tumor growth in mice bearing solid tumor models