Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 4-[(7-chloro-4-quinolinyl)amino]-2-[(diethylamino)methyl]-, hydrochloride (1:2) | 69-44-3 | 99.97% | 1 ML
SDP

Catalog No. 5000411980
Encompass_Preferred

Amodiaquine dihydrochloride is a 4-aminoquinoline antimalarial agent that is orally active. It functions as a potent inhibitor of histamine N-methyltransferase with a Ki of 18.6 nM, and also acts as a Nurr1 agonist, binding specifically to its ligand-binding domain with an EC50 of approximately 20 μM. This compound also demonstrates anti-inflammatory effects and is for research use only.

  • Purity: 99.97%
  • Molecular weight: 428.78
  • Formula: C20H24Cl3N3O
  • CAS number: 69-44-3
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Solubility: Greater than or equal to 125 mg/mL in DMSO (requires sonication), greater than or equal to 50 mg/mL in water (requires sonication)

Catalog No. 50-004-11980 Supplier Medchemexpress LLC Supplier No. HYB1322B10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amodiaquine dihydrochloride is a 4-aminoquinoline antimalarial agent that is orally active. It functions as a potent inhibitor of histamine N-methyltransferase with a Ki of 18.6 nM, and also acts as a Nurr1 agonist, binding specifically to its ligand-binding domain with an EC50 of approximately 20 μM. This compound also demonstrates anti-inflammatory effects and is for research use only.

  • Purity: 99.97%
  • Molecular weight: 428.78
  • Formula: C20H24Cl3N3O
  • CAS number: 69-44-3
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Solubility: Greater than or equal to 125 mg/mL in DMSO (requires sonication), greater than or equal to 50 mg/mL in water (requires sonication)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.