Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 4-amino-3-methyl- | 2835-99-6 | ≥98.0% | 123.15 | 25 G
SDP

Catalog No. 5000452442
Encompass_Preferred

4-Amino-m-cresol inhibits soil urease activity. It is for research use only.

  • Inhibits soil urease activity
  • Appearance: Solid
  • Color: White to off-white
  • Purity: ≥98.0%
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Catalog No. 50-004-52442 Supplier Medchemexpress LLC Supplier No. 25GHYW012824
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Amino-m-cresol inhibits soil urease activity. It is for research use only.

  • Inhibits soil urease activity
  • Appearance: Solid
  • Color: White to off-white
  • Purity: ≥98.0%
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.