Learn More
Medchemexpress LLC Phenol, 4-(aminomethyl)-2-methoxy-, hydrochloride | 7149-10-2 | MFCD00012864 | 98.3% | 189.64 g/mol | C8H12ClNO2 | 250 G
SDP
Catalog No. 5000302565
Encompass_Preferred
Vanillylamine hydrochloride is the hydrochloride salt of vanillylamine supplied as a white to off-white solid for laboratory use. It serves as a biochemical reagent and organic intermediate in synthesis and research involving vanillyl derivatives and capsaicinoid-related studies.
  • White to off-white solid with high purity suitable for research applications.
  • High purity confirmed by LCMS (≈98.3%).
  • Provides a stable hydrochloride salt form for reliable handling and storage.
  • Useful intermediate for synthesis of vanillyl and capsaicinoid derivatives.
  • Associated with standard chemical identifiers for traceability, including CAS 7149-10-2.
Catalog No. 50-003-02565 Supplier Medchemexpress LLC Supplier No. 250GHYW015211
Vanillylamine hydrochloride is the hydrochloride salt of vanillylamine supplied as a white to off-white solid for laboratory use. It serves as a biochemical reagent and organic intermediate in synthesis and research involving vanillyl derivatives and capsaicinoid-related studies.
  • White to off-white solid with high purity suitable for research applications.
  • High purity confirmed by LCMS (≈98.3%).
  • Provides a stable hydrochloride salt form for reliable handling and storage.
  • Useful intermediate for synthesis of vanillyl and capsaicinoid derivatives.
  • Associated with standard chemical identifiers for traceability, including CAS 7149-10-2.