Learn More
Medchemexpress LLC Phenol, 4-(aminomethyl)-2-methoxy-, hydrochloride | 7149-10-2 | MFCD00012864 | 99.8% | 189.64 g/mol | C8H12ClNO2 | 100 G
SDP
Catalog No. 5000302543
Encompass_Preferred
Vanillylamine hydrochloride is the hydrochloride salt of vanillylamine, a small-molecule biochemical reagent used as an intermediate in capsaicinoid synthesis and for life-science research. It is typically supplied as an off-white to pale powder and is commonly used in organic synthesis and biochemical studies.
  • High purity suitable for research applications.
  • Water-soluble hydrochloride salt for easy handling.
  • Useful intermediate in capsaicinoid and vanilloid chemistry.
  • Available in multiple pack sizes to support different scales.
  • Batch-specific certificate of analysis provided for traceability.
Catalog No. 50-003-02543 Supplier Medchemexpress LLC Supplier No. HYW015211100G
Vanillylamine hydrochloride is the hydrochloride salt of vanillylamine, a small-molecule biochemical reagent used as an intermediate in capsaicinoid synthesis and for life-science research. It is typically supplied as an off-white to pale powder and is commonly used in organic synthesis and biochemical studies.
  • High purity suitable for research applications.
  • Water-soluble hydrochloride salt for easy handling.
  • Useful intermediate in capsaicinoid and vanilloid chemistry.
  • Available in multiple pack sizes to support different scales.
  • Batch-specific certificate of analysis provided for traceability.