Learn More
Medchemexpress LLC Phenol, 4-(aminomethyl)- | 696-60-6 | 123.16 | 1 G
SDP
Catalog No. 5000445674
Encompass_Preferred
4-Hydroxybenzylamine is a natural biogenic amine. It can be produced through the breakdown metabolism of glucosinolates, with glutamine serving as the non-classical ammonia donor. It can be utilized for the study of plant amine biosynthesis.
  • Natural biogenic amine
  • Produced through the breakdown metabolism of glucosinolates
  • Can be utilized for the study of plant amine biosynthesis
  • Appears as a solid
  • For research use only
Catalog No. 50-004-45674 Supplier Medchemexpress LLC Supplier No. HYW0040781G
4-Hydroxybenzylamine is a natural biogenic amine. It can be produced through the breakdown metabolism of glucosinolates, with glutamine serving as the non-classical ammonia donor. It can be utilized for the study of plant amine biosynthesis.
  • Natural biogenic amine
  • Produced through the breakdown metabolism of glucosinolates
  • Can be utilized for the study of plant amine biosynthesis
  • Appears as a solid
  • For research use only