Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 4-(aminomethyl)- | 696-60-6 | 699652 | 123.16 | 5 G
SDP

Catalog No. 5000449254
Encompass_Preferred

4-Hydroxybenzylamine is a natural biogenic amine that can be produced through the breakdown metabolism of glucosinolates, with glutamine serving as a non-classical ammonia donor. It can be utilized for the study of plant amine biosynthesis.

  • Natural biogenic amine
  • Produced through glucosinolate breakdown metabolism
  • Useful for plant amine biosynthesis studies
  • Purity: 99.8%
  • Appearance: white to light brown solid
  • Molecular weight: 123.16
  • CAS number: 696-60-6
  • Molecular formula: C7H9NO

Catalog No. 50-004-49254 Supplier Medchemexpress LLC Supplier No. HYW0040785G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroxybenzylamine is a natural biogenic amine that can be produced through the breakdown metabolism of glucosinolates, with glutamine serving as a non-classical ammonia donor. It can be utilized for the study of plant amine biosynthesis.

  • Natural biogenic amine
  • Produced through glucosinolate breakdown metabolism
  • Useful for plant amine biosynthesis studies
  • Purity: 99.8%
  • Appearance: white to light brown solid
  • Molecular weight: 123.16
  • CAS number: 696-60-6
  • Molecular formula: C7H9NO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.