Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 4-ethoxy- | 622-62-8 | MFCD00002334 | 99.9% | 138.16 g/mol | C8H10O2 | 100 G
SDP

Catalog No. 5000300327
Encompass_Preferred

4-Ethoxyphenol (phenol, 4-ethoxy-, CAS 622-62-8) is a small aromatic phenol used as a research reagent in analytical and biological studies. Supplied as a powder, it has molecular formula C8H10O2 and molecular weight 138.16 g/mol. The substance is intended for research use only; consult the certificate of analysis for batch-specific purity and the SDS for safe handling and storage instructions.

  • High purity suitable for analytical applications.
  • Provided as a stable powder for easy handling and storage.
  • Characterized by molecular formula C8H10O2 and MW 138.16 g/mol.
  • Storage recommendations: powder -20°C (long term) or 4°C (short term).
  • Purity verified by HPLC; consult the COA for batch-specific value.

Catalog No. 50-003-00327 Supplier Medchemexpress LLC Supplier No. HYW015786100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Ethoxyphenol (phenol, 4-ethoxy-, CAS 622-62-8) is a small aromatic phenol used as a research reagent in analytical and biological studies. Supplied as a powder, it has molecular formula C8H10O2 and molecular weight 138.16 g/mol. The substance is intended for research use only; consult the certificate of analysis for batch-specific purity and the SDS for safe handling and storage instructions.

  • High purity suitable for analytical applications.
  • Provided as a stable powder for easy handling and storage.
  • Characterized by molecular formula C8H10O2 and MW 138.16 g/mol.
  • Storage recommendations: powder -20°C (long term) or 4°C (short term).
  • Purity verified by HPLC; consult the COA for batch-specific value.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.