Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 5-amino-2,4-difluoro- | 113512-71-3 | 145.11 | 10 G
SDP

Catalog No. 5000460257
Encompass_Preferred

Phenol, 5-amino-2,4-difluoro-, with CAS Number 113512-71-3, is a chemical compound primarily used as a drug intermediate. It boasts a purity of 97.63% and a molecular weight of 145.11. For optimal stability, the solid form should be stored at 4°C, protected from light. When prepared in solvent, it can be stored at -80°C for up to 6 months or at -20°C for 1 month, also under light protection.

  • Chemical name: Phenol, 5-amino-2,4-difluoro-
  • CAS Number: 113512-71-3
  • Used as a drug intermediate
  • Purity: 97.6%
  • Molecular weight: 145.11
  • Solid storage: 4°C, protect from light
  • Solvent storage: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Appearance: Light brown to brown solid

Catalog No. 50-004-60257 Supplier Medchemexpress LLC Supplier No. HYW00727210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenol, 5-amino-2,4-difluoro-, with CAS Number 113512-71-3, is a chemical compound primarily used as a drug intermediate. It boasts a purity of 97.63% and a molecular weight of 145.11. For optimal stability, the solid form should be stored at 4°C, protected from light. When prepared in solvent, it can be stored at -80°C for up to 6 months or at -20°C for 1 month, also under light protection.

  • Chemical name: Phenol, 5-amino-2,4-difluoro-
  • CAS Number: 113512-71-3
  • Used as a drug intermediate
  • Purity: 97.6%
  • Molecular weight: 145.11
  • Solid storage: 4°C, protect from light
  • Solvent storage: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Appearance: Light brown to brown solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.