Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenol, 5-amino-2,4-difluoro- | 113512-71-3 | 145.11 | 5 G
SDP

Catalog No. 5000460392
Encompass_Preferred

Phenol, 5-amino-2,4-difluoro- is a chemical compound with a molecular weight of 145.11 and a purity of 97.63%. It appears as a light brown to brown solid. This product has been tested and complies with the given specifications.

  • Molecular formula: C6H5F2NO
  • Appearance: Light brown to brown solid
  • Purity: 97.63%
  • Storage: 4°C, protect from light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Catalog No. 50-004-60392 Supplier Medchemexpress LLC Supplier No. HYW0072725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenol, 5-amino-2,4-difluoro- is a chemical compound with a molecular weight of 145.11 and a purity of 97.63%. It appears as a light brown to brown solid. This product has been tested and complies with the given specifications.

  • Molecular formula: C6H5F2NO
  • Appearance: Light brown to brown solid
  • Purity: 97.63%
  • Storage: 4°C, protect from light
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.