Learn More
Medchemexpress LLC Phenol, 5-chloro-2-(4,6-dichlorophenoxy-2,3,5-d3)- | 1020719-98-5 | 98.2% | 1 MG
SDP
Catalog No. 5000413481
Encompass_Preferred
Triclosan-d3 is the deuterium labeled Triclosan. Triclosan is an antibacterial and antifungal agent found in consumer products, including soaps, detergents, toys, and surgical cleaning treatments.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process.
  • Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13481 Supplier Medchemexpress LLC Supplier No. HYB1119S1MG
Triclosan-d3 is the deuterium labeled Triclosan. Triclosan is an antibacterial and antifungal agent found in consumer products, including soaps, detergents, toys, and surgical cleaning treatments.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process.
  • Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.