Learn More
Medchemexpress LLC Phenol-amido-C1-PEG3-N3 | 1096439-18-7 | 99.9% | 10 MG
SDP
Catalog No. 5000338771
Encompass_Preferred
Phenol-amido-C1-PEG3-N3 (PROTAC Linker 21) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing Alkyne groups. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing alkyne groups
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-38771 Supplier Medchemexpress LLC Supplier No. HY12883510MG
Phenol-amido-C1-PEG3-N3 (PROTAC Linker 21) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing Alkyne groups. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing alkyne groups
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups