Learn More
Sigma Aldrich Fine Chemicals Biosciences PHENOL:CHLOROFORM:ISOAMYL100ML
Catalog No. NC2086495
Encompass
Phenol:Chloroform:Isoamyl Alcohol 25:24:1, Saturated with 10mM Tris, pH 8.0, 1mM EDTA 100mL
This item is not returnable. View return policy
Catalog No. NC2086495 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P3803100ML
Phenol:Chloroform:Isoamyl Alcohol 25:24:1, Saturated with 10mM Tris, pH 8.0, 1mM EDTA 100mL