Learn More
Medchemexpress LLC Phenol, dimethyl-, 1,1',1''-phosphate | 25155-23-1 | 98.1% | 500 MG
SDP
Catalog No. 5000230246
Encompass_Preferred
Trixylyl phosphate (CAS 25155-23-1) is an aromatic triaryl phosphate ester used as a plasticizer and flame-retardant additive in industrial and consumer formulations, including plastics and hydraulic fluids. Supplied for research and formulation use with accompanying certificate of analysis and safety data sheet.
  • Aromatic triaryl phosphate ester used as plasticizer and flame retardant.
  • Common applications include plastics, hydraulic fluids, and flame-retardant formulations.
  • High chemical purity, typically about 98.1%.
  • Molecular formula C24H27O4P; molecular weight ≈ 410.4 g/mol.
  • Documentation available: certificate of analysis and safety data sheet.
Catalog No. 50-002-30246 Supplier Medchemexpress LLC Supplier No. HY122519500MG
Trixylyl phosphate (CAS 25155-23-1) is an aromatic triaryl phosphate ester used as a plasticizer and flame-retardant additive in industrial and consumer formulations, including plastics and hydraulic fluids. Supplied for research and formulation use with accompanying certificate of analysis and safety data sheet.
  • Aromatic triaryl phosphate ester used as plasticizer and flame retardant.
  • Common applications include plastics, hydraulic fluids, and flame-retardant formulations.
  • High chemical purity, typically about 98.1%.
  • Molecular formula C24H27O4P; molecular weight ≈ 410.4 g/mol.
  • Documentation available: certificate of analysis and safety data sheet.