Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Fused Crystal, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18605076 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
6 x 500 g
25 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-076 25 kg Amber Glass Bottle
18-605-070 25 g Amber Glass Bottle
18-605-072 500 g Amber Glass Bottle
18-605-074 1 kg Amber Glass Bottle
18-605-073 6 x 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-605-076 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PH12025KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

PH120, 108-95-2, C6H6O

Spectrum™ Chemical Phenol, Fused Crystal, USP is extracted from petroleum and mainly used to produce precursors of plastics and is also used as a building block to synthetically produce a wide range of pharmaceutical products and drugs such as aspirin and oral analgesic among them. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Color Colorless
Quantity 25 kg
Physical Form Fused Crystal
Percent Purity 99 to 100.5%
Grade USP
Chemical Name or Material Phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.