Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Fused Crystal, White, Ultrapure Bioreagent, J.T. Baker™

Catalog No. p-5391245 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Steel Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-841 1 kg Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-841 Supplier Avantor Supplier No. 405602

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Extraction of Nucleic Acids and Molecular Biology Applications

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Color White
Flash Point 79.00°C
Packaging Steel Can
Quantity 1 kg
Specific Gravity 1L = 1.06kg
Formula Weight 94.11
Grade Ultrapure Bioreagent
Physical Form Fused Crystals
Chemical Name or Material Phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.