Learn More
LaMarKa Chemical Company Phenolphthalein Indicator 10gpl (1%W/V)(in Methanol) ASTM E200, 1l
Catalog No. 504400839
Encompass_Preferred
Phenolphthalein Indicator 10gpl (1%W/V)(in Methanol) ASTM E200, 1l Plastic Bottle
Catalog No. 50-440-0839 Supplier LaMarKa Chemical Company Supplier No. 0000686816750101
Add to Cart
Add to Cart
Phenolphthalein Indicator 10gpl (1%W/V)(in Methanol) ASTM E200, 1l Plastic Bottle