Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Liquefied, 89%, Spectrum™ Chemical
SDP

Catalog No. p-7166013 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
2.5 L
4 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-224 120 mL Amber Glass Bottle
18-612-227 500 mL Amber Glass Bottle
18-612-225 2.5 L Amber Glass Bottle
18-612-258 4 L Amber Glass Bottle
18-612-226 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-612-224 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P1070120ML
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-95-2 , 7732-18-5

Specifications

Packaging Amber Glass Bottle
Quantity 120 mL
Residue after Evaporation 0.0005
Percent Purity 0.89
Grade Reagent
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.