Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Loose Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18605086 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
1 kg
6 x 125 g
6 x 500 g
4 x 1 kg
45 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-086 45 kg Amber Glass Bottle
18-605-077 125 g Amber Glass Bottle
18-605-079 500 g Amber Glass Bottle
18-605-078 6 x 125 g Amber Glass Bottle
18-605-081 1 kg Amber Glass Bottle
18-605-080 6 x 500 g Amber Glass Bottle
18-605-082 4 x 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-605-086 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P106045KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P1060, 108-95-2, C6H6O

Spectrum™ Chemical Phenol, Loose Crystal, Reagent, ACS is a white crystalline, aromatic organic compound that is volatile. It's extracted from petroleum and mainly used as a synthetic building block to produce a wide range of pharmaceutical products and drugs such as aspirin and oral analgesics. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Color Colorless to White
Quantity 45 kg
Physical Form Loose Crystals
Percent Purity 99%
Grade ACS
Chemical Name or Material Phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.