Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, MP Biomedicals™

Catalog No. MP04818048 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP04818048 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP04818048 Supplier MP Biomedicals, Inc Supplier No. 04818048
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenol, Ultra Pure Grade, White Solid

  • Phenol is useful for extraction of viral, plasmid and phage DNAs, and to solubilize and dissociate proteins.
  • It is used as a general disinfectant, either in solution or mixed with slaked lime, etc., for toilets, stables, cesspools, floors, drains, etc.; for the manufacture of colorless or light-colored artificial resins, many medical and industrial organic compounds and dyes; as a reagent in chemical analysis.
  • Aqueous solutions are used as topical anesthetic; topical antiseptic; topical antipruritic. Phenol is used in DNA extractions for removing protein.
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-95-2
Melting Point 182°C (literature)
Color White
Density 1.071g/mL at 25°C (Lit.)
Boiling Point 182°C (Literature)
Flash Point 174.2°F/79°C (Lit.)
Molecular Formula C6H6O
Quantity 500 g
MDL Number MFCD00002143
UN Number r
Residue after Evaporation <0.05%
Beilstein 969616
Synonym Hydroxybenzene, Carbolic acid, Phenic acid, Phenylic acid
Solubility Information Soluble in aqueous solution (5% - clear, colorless solution).
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1
IUPAC Name phenol
Molecular Weight (g/mol) 94.11
Vapor Density 3.24 (vs air)(Lit.)
Vapor Pressure 0.09 psi (55°C); 0.36 mmHg (20°C)(Lit.)
Percent Purity 0.999
Physical Form Solid
Assay Ultra Puregrade
Show More Show Less
storageNote1 Store at Room Temperature (15-30°C), store under nitrogen.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.