Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019620 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9620 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9620 Supplier MilliporeSigma Supelco Supplier No. PHR10471G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenol is a derivative of benzene and is majorly used in the manufacture of bisphenol A, phenolic resins and caprolactam. Its other applications include the manufacture of variety of compounds in pharma industry, dyes, explosives, rubber and agriculture.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-95-2
Vapor Pressure 0.36 mmHg (20°C)
Vapor Density 3.24 (vs. air)
Density 1.071 g/mL (at 25°C)
Linear Formula C6H5OH
MDL Number MFCD00002143
Melting Point 40°C to 42°C
Boiling Point 182°C
Synonym Hydroxybenzene
Quantity 1 g
Formula Weight 94.11
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.