Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Purified, Liquefied, Reagents

Catalog No. 01337141
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-141 100 mL
01-337-140 1 L
01-337-139 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-141 Supplier Reagents Holdings Llc Supplier No. C2269500100C
Add to Cart
Add to Cart

Phenol, Purified, Liquefied, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

Synonym Liquified phenol

Specifications

CAS 7732-18-5
Boiling Range 182°C
Color Colorless to Slightly Pink
pH 3.5 to 4.2
Boiling Point 182°C
Packaging Glass Bottle
Molecular Formula H2O
Quantity 100 mL
Specific Gravity 1.06
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Grade Pure
Physical Form Liquid
Chemical Name or Material Phenol, Purified, Liquefied
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.